missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human STOML3 (aa 1-26) Control Fragment Recombinant Protein

Numéro de catalogue. RP90230
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
RP90230 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Numéro de catalogue. RP90230 Fournisseur Invitrogen™ Code fournisseur RP90230
Il en reste null

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (50%), Rat (50%). This recombinant protein control fragment may be used for blocking experiments. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

STOML3 is required for the function of many mechanoreceptors. It modulates mechanotransduction channels and acid-sensing ion channels (ASIC) proteins. The protein potentiates PIEZO1 and PIEZO2 function by increasing their sensitivity to mechanical stimulations.

Spécifications

Accession Number Q8TAV4
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 161003
Name Human STOML3 (aa 1-26) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias Epb7.2 L; EPB72-like 3; erythrocyte band 7 integral membrane protein; protein 7.2 B; SLP3; SLP-3; sro; stomatin (Epb7.2)-like 3; stomatin (EPB72)-like 3; stomatin like 3; stomatin related olfactory protein SRO; stomatin-like protein 3; Stomatin-related olfactory protein; STOML3
Common Name STOML3
Gene Symbol Stoml3
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence MDSRVSSPEKQDKENFVGVNNKRLGV
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Afficher plus de résultats Afficher moins de résultats