missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human SMPD1 ELISA Kit
ELISA
$890.00 - $4,420.00
Spécifications
| Accession Number | Acid sphingomyelinase; Asm; aSMase; A-SMase; NPD; Smpd1; sphingomyelin phosphodiesterase; sphingomyelin phosphodiesterase 1; sphingomyelin phosphodiesterase 1, acid lysosomal; Zn-SMase |
|---|---|
| Assay Range | 1.22 to 300 ng/mL |
| Assay Sensitivity | 1.22 ng/mL |
| Conjugate | HRP |
| Product Type | ELISA |
| Numéro de catalogue | Numéro du manufacturier. | Quantity | Prix | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Numéro de catalogue | Numéro du manufacturier. | Quantity | Prix | Quantity & Availability | |||||
EH431RBX5
|
Invitrogen™
EH431RBX5 |
5 x 96 Tests |
chaque for $4,420.00
|
|
Connectez-vous ou enregistrez-vous pour vérifier votre prix et la disponibilité.
|
||||
EH431RB
|
Invitrogen™
EH431RB |
96 Tests |
chaque for $890.00
|
|
Connectez-vous ou enregistrez-vous pour vérifier votre prix et la disponibilité.
|
||||
Description
Human SMPD1 quantitates human SMPD1 in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant human SMPD1.
The protein encoded by this gene is a lysosomal acid sphingomyelinase that converts sphingomyelin to ceramide. The encoded protein also has phospholipase C activity. Defects in this gene are a cause of Niemann-Pick disease type A (NPA) and Niemann-Pick disease type B (NPB). Three transcript variants encoding two different isoforms have been found for this gene.Spécifications
| Acid sphingomyelinase; Asm; aSMase; A-SMase; NPD; Smpd1; sphingomyelin phosphodiesterase; sphingomyelin phosphodiesterase 1; sphingomyelin phosphodiesterase 1, acid lysosomal; Zn-SMase | |
| 1.22 ng/mL | |
| ELISA | |
| Colorimetric Microplate Reader | |
| Smpd1 | |
| <10 | |
| HRP | |
| Plasma 50 μL, Serum 50 μL, Supernatant 50 μL | |
| ASM | |
| 4 hrs 45 mins |
| 1.22 to 300 ng/mL | |
| HRP | |
| Plasma, Serum, Supernatant | |
| 6609 | |
| <12% | |
| Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers | |
| RUO | |
| 2°C to 8°C | |
| 1 hrs 20 mins |