missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human SARS-CoV-2 Spike (Trimer) Ig Total ELISA Kit

Numéro de catalogue. BMS2323TWO
Encompass
Modifier l'affichage
Click to view available options
Quantity:
2 x 96 Tests
96 Tests
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
BMS2323TWO 2 x 96 Tests
BMS2323 96 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Numéro de catalogue. BMS2323TWO Fournisseur Invitrogen™ Code fournisseur BMS2323TWO
Il en reste null

ELISA

The Human SARS-CoV-2 Spike (Trimer) Ig Total ELISA is a serology assay that measures and quantitates immunoglobulin (Ig) antibodies against SARS-CoV-2 Spike (Trimer) in human serum or plasma. Principle of the method The Human SARS-CoV-2 Spike (Trimer) Ig Total ELISA (enzyme-linked immunosorbent assay) is designed to measure the amount of Ig antibodies bound to SARS-CoV-2 Spike (Trimer). A trimerized Spike protein is pre-coated in the wells of the supplied microplate. Samples, controls, including a high control that can be used as a standard are then added into these wells and bind to the immobilized (capture) Spike protein. The wells are washed and Ig antibodies conjugated to HRP are added and will bind to any captured antibodies. The Ig Total antibody recognizes IgG, IgM and IgA. The wells are washed and a substrate solution is added that reacts with the enzyme complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of antibody present in the original specimen.

It has recently been shown that SARS (severe acute respiratory syndrome) is caused by a human coronavirus. Human coronaviruses are the major cause of upper respiratory tract illness, such as the common cold, in humans. Coronaviruses are positive-stranded RNA viruses, featuring the largest viral RNA genomes known to date (27-31 kb). The first step in coronavirus infection is binding of the viral spike protein, a 139-kDa protein, to certain receptors on host cells. The spike protein is the main surface antigen of the coronavirus. The glycosylated spike protein (as well as the nucleocapsid protein) can be detected in infected cell culture supernatants with antisera from SARS patients.

Spécifications

Accession Number Coronavirus; COVID; Covid 19; COVID19; COVID-19; SARS; SARS Coronavirus; SARS CoV; SARS CoV2; SARS CoV-2; SARS Spike Protein; SARS-CoV-2
Assay Range 62.5 to 4000 Units/mL
Assay Sensitivity <62.5 U/mL
Conjugate HRP
Product Type ELISA
Sample Type Plasma, Serum
For Use With (Equipment) Colorimetric Microplate Reader
Interassay CV 5%
Intraassay CV 4.3
Kit Contents Pre-coated 96 well plate, High Control (standard), Calibrator (medium control), Low Control, Anti-Ig Total - HRP conjugate, Assay Buffer concentrate, Wash Buffer concentrate, Substrate solution, Stop Solution, Adhesive Plate Covers
Label or Dye HRP
Quantity 2 x 96 Tests
Regulatory Status RUO
Sample Volume Plasma 5 μL, Serum 5 μL
Storage Requirements 2°C to 8°C
Target SARS-CoV-2 Spike (trimer) Protein
Test Time 30 mins
Total Assay Time 1 hrs 15 mins
Afficher plus de résultats Afficher moins de résultats