missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human OX40L ELISA Kit
ELISA
Supplier: Invitrogen™ EH356RB
Description
Human OX40L quantitates human OX40L in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant human OX40L.
The tumor necrosis factor superfamily member TNFSF4 is a type II membrane bound, non-covalently linked homotrimeric protein. It is expressed on antigen presenting cells, such as dendritic cells and activated B-cells, and also on other cells such as vascular endothelial cells, mast cells, and natural killer cells. TNFSF4 signals specifically through the TNFRSF4 receptor, is expressed predominantly on CD4+T cells but also on certain activated CD8+T cells. TNFRSF4/TNFSF4 functions as a costimulatory signal, which is required for a productive interaction between antigen presenting cells and their target T-cells. It enhances cell proliferation and survival, and increases expression of RANTES, IL-2, IL-3, and IFNgamma. TNFRSF4/TNFSF4 signaling plays an important role in immunotolerance.Identifiants chimiques
| Ath1; Ath-1; atherosclerosis 1; CD134 ligand; CD134L; CD252; Glycoprotein Gp34; GP34; O x 40 antigen ligand; O x 40 ligand; O x 40 L; OX-40 L; O x 4 OL; TAX transcriptionally-activated glycoprotein 1; tax-transcriptionally activated glycoprotein 1 (34 kD); tax-transcriptionally activated glycoprotein 1 ligand; Tnfsf4; TNLG2B; tumor necrosis factor (ligand) superfamily member 4; tumor necrosis factor (ligand) superfamily, member 4; tumor necrosis factor ligand 2 B; tumor necrosis factor ligand superfamily member 4; tumor necrosis factor superfamily member 4; Txgp1; Txgp1l | |
| 61 pg/mL | |
| ELISA | |
| Colorimetric Microplate Reader | |
| Tnfsf4 | |
| <10 | |
| HRP | |
| Plasma 50 μL, Serum 50 μL, Supernatant 50 μL | |
| OX40L (CD252) | |
| 4 hrs 45 mins |
| 61 to 15,000 pg/mL | |
| HRP | |
| Plasma, Serum, Supernatant | |
| 7292 | |
| <12% | |
| Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers | |
| RUO | |
| 2°C to 8°C | |
| 1 hrs 20 mins |
Spécifications
| Ath1; Ath-1; atherosclerosis 1; CD134 ligand; CD134L; CD252; Glycoprotein Gp34; GP34; O x 40 antigen ligand; O x 40 ligand; O x 40 L; OX-40 L; O x 4 OL; TAX transcriptionally-activated glycoprotein 1; tax-transcriptionally activated glycoprotein 1 (34 kD); tax-transcriptionally activated glycoprotein 1 ligand; Tnfsf4; TNLG2B; tumor necrosis factor (ligand) superfamily member 4; tumor necrosis factor (ligand) superfamily, member 4; tumor necrosis factor ligand 2 B; tumor necrosis factor ligand superfamily member 4; tumor necrosis factor superfamily member 4; Txgp1; Txgp1l | |
| 61 pg/mL | |
| ELISA | |
| Colorimetric Microplate Reader | |
| Tnfsf4 | |
| <10 | |
| HRP | |
| RUO | |
| 2°C to 8°C | |
| 1 hrs 20 mins |
| 61 to 15,000 pg/mL | |
| HRP | |
| Plasma, Serum, Supernatant | |
| 7292 | |
| <12% | |
| Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers | |
| 96 Tests | |
| Plasma 50 μL, Serum 50 μL, Supernatant 50 μL | |
| OX40L (CD252) | |
| 4 hrs 45 mins |