missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human Meprin alpha ELISA Kit
ELISA
Supplier: Invitrogen™ EH321RB
Description
Human Meprin alpha quantitates human Meprin alpha in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant human Meprin alpha.
MEP1A (Meprin A Subunit Alpha) is a Protein Coding gene. Among its related pathways are Collagen chain trimerization. Gene Ontology (GO) annotations related to this gene include metalloendopeptidase activity and metallopeptidase activity.Identifiants chimiques
| AI098089; AW107200; bA268F1.1 (meprin A alpha (PABA peptide hydrolase)); E-24.18; Endopeptidase-2; Endopeptidase-24.18 subunit alpha; Mep1; MEP-1; Mep1a; Mep-1 A; meprin 1 alpha; meprin A alpha; meprin A alpha-subunit; Meprin A subunit alpha; meprin A, alpha (PABA peptide hydrolase); meprin alpha; N-benzoyl-L-tyrosyl-P-amino-benzoic acid hydrolase subunit alpha; PABA peptide hydrolase; PPH alpha; PPHA | |
| 2 ng/mL | |
| ELISA | |
| Colorimetric Microplate Reader | |
| MEP1A | |
| <10 | |
| HRP | |
| Plasma 50 μL, Serum 50 μL, Supernatant 50 μL | |
| MEP1A | |
| 4 hrs 45 mins |
| 2 to 400 ng/mL | |
| HRP | |
| Plasma, Serum, Supernatant | |
| 4224 | |
| <12% | |
| Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers | |
| RUO | |
| 2°C to 8°C | |
| 1 hrs 20 mins |
Spécifications
| AI098089; AW107200; bA268F1.1 (meprin A alpha (PABA peptide hydrolase)); E-24.18; Endopeptidase-2; Endopeptidase-24.18 subunit alpha; Mep1; MEP-1; Mep1a; Mep-1 A; meprin 1 alpha; meprin A alpha; meprin A alpha-subunit; Meprin A subunit alpha; meprin A, alpha (PABA peptide hydrolase); meprin alpha; N-benzoyl-L-tyrosyl-P-amino-benzoic acid hydrolase subunit alpha; PABA peptide hydrolase; PPH alpha; PPHA | |
| 2 ng/mL | |
| ELISA | |
| Colorimetric Microplate Reader | |
| MEP1A | |
| <10 | |
| HRP | |
| RUO | |
| 2°C to 8°C | |
| 1 hrs 20 mins |
| 2 to 400 ng/mL | |
| HRP | |
| Plasma, Serum, Supernatant | |
| 4224 | |
| <12% | |
| Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers | |
| 96 Tests | |
| Plasma 50 μL, Serum 50 μL, Supernatant 50 μL | |
| MEP1A | |
| 4 hrs 45 mins |