missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human MEL18 (aa 244-286) Control Fragment Recombinant Protein

Numéro de catalogue. RP107245
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
RP107245 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Numéro de catalogue. RP107245 Fournisseur Invitrogen™ Code fournisseur RP107245
Il en reste null

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (100%), Rat (100%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-66592 (PA5-66592. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

The protein encoded by this gene contains a RING finger motif and is similar to the polycomb group (PcG) gene products. PcG gene products form complexes via protein-protein interaction and maintain the transcription repression of genes involved in embryogenesis, cell cycles, and tumorigenesis. This protein was shown to act as a negative regulator of transcription and has tumor suppressor activity. The expression of this gene was detected in various tumor cells, but is limited in neural organs in normal tissues. Knockout studies in mice suggested that this protein may negatively regulate the expression of different cytokines, chemokines, and chemokine receptors, and thus plays an important role in lymphocyte differentiation and migration, as well as in immune responses.

Spécifications

Accession Number P35227
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 7703
Name Human MEL18 (aa 244-286) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias DNA-binding protein Mel-18; Mel18; Mel-18; Melanoma nuclear protein 18; MGC10545; Pcgf2; polycomb group ring finger 2; polycomb group RING finger protein 2; RING finger protein 110; RNF110; Zfp144; Zfp-144; Zinc finger protein 144; zinc finger protein 144 (Mel-18) (human homolog); ZNF144
Common Name MEL18
Gene Symbol PCGF2
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence PSEGTNTSGASECESVSDKAPSPATLPATSSSLPSPATPSHGS
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Afficher plus de résultats Afficher moins de résultats