missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human MD-2 ELISA Kit
ELISA
$890.00 - $4,420.00
Spécifications
| Accession Number | ESOP1; ESOP-1; LPS co-receptor MD-2; LY96; ly-96; lymphocyte antigen 96; MD2; MD-2; myeloid differentiation factor-2; myeloid differentiation protein-2; protein MD-2 |
|---|---|
| Assay Range | 1.6 to 400 ng/mL |
| Assay Sensitivity | 1.6 ng/mL |
| Conjugate | HRP |
| Product Type | ELISA |
| Numéro de catalogue | Numéro du manufacturier. | Quantity | Prix | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Numéro de catalogue | Numéro du manufacturier. | Quantity | Prix | Quantity & Availability | |||||
EH318RBX5
|
Invitrogen™
EH318RBX5 |
5 x 96 Tests |
chaque for $4,420.00
|
|
Connectez-vous ou enregistrez-vous pour vérifier votre prix et la disponibilité.
|
||||
EH318RB
|
Invitrogen™
EH318RB |
96 Tests |
chaque for $890.00
|
|
Connectez-vous ou enregistrez-vous pour vérifier votre prix et la disponibilité.
|
||||
Description
Human MD-2 quantitates human MD-2 in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant human MD-2.
MD-2 is a gene that encodes a protein involved in the recognition of bacterial lipopolysaccharides (LPS) by the immune system. It is located on chromosome 8 and is part of a larger family of genes that encode for similar proteins. The MD-2 protein forms a complex with Toll-like receptor 4 (TLR4) to recognize and respond to LPS, triggering an immune response. Mutations or variations in the MD-2 gene have been associated with various diseases and conditions, including sepsis and inflammatory disorders.Spécifications
| ESOP1; ESOP-1; LPS co-receptor MD-2; LY96; ly-96; lymphocyte antigen 96; MD2; MD-2; myeloid differentiation factor-2; myeloid differentiation protein-2; protein MD-2 | |
| 1.6 ng/mL | |
| ELISA | |
| Colorimetric Microplate Reader | |
| LY96 | |
| <10 | |
| HRP | |
| Plasma 50 μL, Serum 50 μL, Supernatant 50 μL | |
| MD-2 | |
| 4 hrs 45 mins |
| 1.6 to 400 ng/mL | |
| HRP | |
| Plasma, Serum, Supernatant | |
| 23643 | |
| <12% | |
| Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers | |
| RUO | |
| 2°C to 8°C | |
| 1 hrs 20 mins |