missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human JAM-B ELISA Kit
ELISA
Supplier: Invitrogen™ EH284RB
Description
Human JAM-B quantitates human JAM-B in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant human JAM-B.
Tight junctions represent one mode of cell-to-cell adhesion in epithelial or endothelial cell sheets, forming continuous seals around cells and serving as a physical barrier to prevent solutes and water from passing freely through the paracellular space. The protein encoded by this immunoglobulin superfamily gene member is localized in the tight junctions between high endothelial cells. It acts as an adhesive ligand for interacting with a variety of immune cell types and may play a role in lymphocyte homing to secondary lymphoid organs.Identifiants chimiques
| 1110002N23Rik; 2410030G21Rik; 2410167M24Rik; AU016127; C21orf43; CD322; Jam2; JAM-2; JAMB; JAM-B; JAM-IT/VE-JAM; Jcam2; junction adhesion molecule 2; junction adhesion molecule B; junction cell adhesion molecule 2; junctional adhesion molecule 2; Junctional adhesion molecule B; PRO245; RP23-253E17.2; UNQ219/PRO245; Vascular endothelial junction-associated molecule; VEJAM; VE-JAM | |
| 0.068 ng/mL | |
| ELISA | |
| Colorimetric Microplate Reader | |
| JAM2 | |
| <10 | |
| HRP | |
| Plasma 50 μL, Serum 50 μL, Supernatant 50 μL | |
| JAM2 | |
| 4 hrs 45 mins |
| 0.068 to 50 ng/mL | |
| HRP | |
| Plasma, Serum, Supernatant | |
| 58494 | |
| <12% | |
| Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers | |
| RUO | |
| 2°C to 8°C | |
| 1 hrs 20 mins |
Spécifications
| 1110002N23Rik; 2410030G21Rik; 2410167M24Rik; AU016127; C21orf43; CD322; Jam2; JAM-2; JAMB; JAM-B; JAM-IT/VE-JAM; Jcam2; junction adhesion molecule 2; junction adhesion molecule B; junction cell adhesion molecule 2; junctional adhesion molecule 2; Junctional adhesion molecule B; PRO245; RP23-253E17.2; UNQ219/PRO245; Vascular endothelial junction-associated molecule; VEJAM; VE-JAM | |
| 0.068 ng/mL | |
| ELISA | |
| Colorimetric Microplate Reader | |
| JAM2 | |
| <10 | |
| HRP | |
| RUO | |
| 2°C to 8°C | |
| 1 hrs 20 mins |
| 0.068 to 50 ng/mL | |
| HRP | |
| Plasma, Serum, Supernatant | |
| 58494 | |
| <12% | |
| Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers | |
| 96 Tests | |
| Plasma 50 μL, Serum 50 μL, Supernatant 50 μL | |
| JAM2 | |
| 4 hrs 45 mins |