missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human IQCD (aa 329-403) Control Fragment Recombinant Protein

Numéro de catalogue. RP100816
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
RP100816 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Numéro de catalogue. RP100816 Fournisseur Invitrogen™ Code fournisseur RP100816
Il en reste null

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (77%), Rat (77%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-61818 (PA5-61818. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

Spécifications

Accession Number Q96DY2
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 115811
Name Human IQCD (aa 329-403) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias 4933433C09Rik; AI662646; CFAP84; Drc10; Dynein regulatory complex protein 10; dynein regulatory complex subunit 10; IQ domain-containing protein D; IQ motif containing D; Iqcd
Common Name IQCD
Gene Symbol IQCD
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence EIENWIQKYDTEMGEKQEELEDLDAVHREEKISLEELRRRHKVLVGEFAQIREEREINSKKRMEAEQEMVRMVRA
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Afficher plus de résultats Afficher moins de résultats