missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human GDPD3 (aa 23-84) Control Fragment Recombinant Protein

Numéro de catalogue. RP98553
Modifier l'affichage
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
RP98553 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Numéro de catalogue. RP98553 Fournisseur Invitrogen™ Code fournisseur RP98553
Il en reste null

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (87%), Rat (87%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-59581 (PA5-59581. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

Spécifications

Accession Number Q7L5L3
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 79153
Name Human GDPD3 (aa 23-84) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias 1110015E22Rik; GDE7; Gdpd3; Glycerophosphodiester phosphodiesterase 7; glycerophosphodiester phosphodiesterase domain containing 3; glycerophosphodiester phosphodiesterase domain-containing protein 3; Lysophospholipase D GDPD3; RGD1308386
Common Name GDPD3
Gene Symbol GDPD3
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence LRRPHLLHTPRAPTFRIRLGAHRGGSGELLENTMEAMENSMAQRSDLLELDCQLTRDRVVVV
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Afficher plus de résultats Afficher moins de résultats