missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gibco™ Human FGF-9 Recombinant Protein, PeproTech®
Description
100-00CC-500UG will be provided as 2 x 250 μg (100-00CC-250UG). Recombinant Human FGF-9 is a 23.2 kDa protein consisting of 206 amino acid residues. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis
The protein encoded by this gene is a member of the fibroblast growth factor (FGF) family. FGF family members possess broad mitogenic and cell survival activities, and are involved in a variety of biological processes, including embryonic development, cell growth, morphogenesis, tissue repair, tumor growth and invasion. This protein was isolated as a secreted factor that exhibits a growth-stimulating effect on cultured glial cells. In nervous system, this protein is produced mainly by neurons and may be important for glial cell development. Expression of the mouse homolog of this gene was found to be dependent on Sonic hedgehog (Shh) signaling. Mice lacking the homolog gene displayed a male-to-female sex reversal phenotype, which suggested a role in testicular embryogenesis.
Spécifications
Spécifications
| Accession Number | P31371 |
| For Use With (Application) | Functional Assay |
| Formulation | protein with no preservative |
| Gene ID (Entrez) | 2254 |
| Molecular Weight (g/mol) | 23.2 kDa |
| Name | Human FGF-9 |
| Quantity | 100 μg |
| Source | E. coli |
| Regulatory Status | RUO |
| Endotoxin Concentration | <1 EU/ μg |
| Afficher plus de résultats |