missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human FGF-17 ELISA Kit
ELISA
Supplier: Invitrogen™ EH186RB
Description
Human FGF-17 quantitates human FGF-17 in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant human FGF-17.
The protein encoded by this gene is a member of the fibroblast growth factor (FGF) family. FGF family members possess broad mitogenic and cell survival activities, and are involved in a variety of biological processes including embryonic development cell growth, morphogenesis, tissue repair, tumor growth and invasion. This gene was shown to be prominently expressed in the cerebellum and cortex. The mouse homolog of this gene was localized to specific sites in the midline structures of the forebrain, the midbrain-hindbrain junction, developing skeleton and developing arteries, which suggests a role in central nervous system, bone and vascular development. This gene was referred to as FGF-13 in reference 2, however, its amino acid sequence and chromosomal localization are identical to FGF17.Identifiants chimiques
| FGF; FGF-13; FGF17; FGF-17; Fibroblast growth factor; Fibroblast growth factor 17; HH20; UNQ161/PRO187 | |
| 0.25 ng/mL | |
| ELISA | |
| Colorimetric Microplate Reader | |
| FGF17 | |
| <10 | |
| HRP | |
| Plasma 50 μL, Serum 50 μL, Supernatant 50 μL | |
| FGF17 | |
| 4 hrs 45 mins |
| 0.25 to 60 ng/mL | |
| HRP | |
| Plasma, Serum, Supernatant | |
| 8822 | |
| <12% | |
| Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers | |
| RUO | |
| 2°C to 8°C | |
| 1 hrs 20 mins |
Spécifications
| FGF; FGF-13; FGF17; FGF-17; Fibroblast growth factor; Fibroblast growth factor 17; HH20; UNQ161/PRO187 | |
| 0.25 ng/mL | |
| ELISA | |
| Colorimetric Microplate Reader | |
| FGF17 | |
| <10 | |
| HRP | |
| RUO | |
| 2°C to 8°C | |
| 1 hrs 20 mins |
| 0.25 to 60 ng/mL | |
| HRP | |
| Plasma, Serum, Supernatant | |
| 8822 | |
| <12% | |
| Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers | |
| 96 Tests | |
| Plasma 50 μL, Serum 50 μL, Supernatant 50 μL | |
| FGF17 | |
| 4 hrs 45 mins |