missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human CCDC107 (aa 25-61) Control Fragment Recombinant Protein

Numéro de catalogue. RP104902
Modifier l'affichage
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
RP104902 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Numéro de catalogue. RP104902 Fournisseur Invitrogen™ Code fournisseur RP104902
Il en reste null

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (54%), Rat (54%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-65619 (PA5-65619. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

This gene encodes a membrane protein which contains a coiled-coil domain in the central region. Multiple alternatively spliced transcript variants encoding different isoforms have been found for this gene.

Spécifications

Accession Number Q8WV48
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 203260
Name Human CCDC107 (aa 25-61) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias CCDC107; coiled-coil domain containing 107; coiled-coil domain-containing protein 107; PSEC0222
Common Name CCDC107
Gene Symbol CCDC107
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence DRANPDLRAHPGNAAHPGSGATEPRRRPPLKDQRERT
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Afficher plus de résultats Afficher moins de résultats