missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human beta-Glucuronidase/GUSB ELISA Kit
ELISA
Supplier: Invitrogen™ EH43RB
Description
Human beta-Glucuronidase/GUSB quantitates human beta-Glucuronidase/GUSB in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant human beta-Glucuronidase/GUSB.
The GUSB gene encodes beta-glucuronidase (EC 3.2.1.31), a lysosomal hydrolase involved in the stepwise degradation of glucuronic acid-containing glycosaminoglycans. It is a tetrameric glycoprotein composed of identical subunits. The GUSB gene is mutated in mucopolysaccharidosis type VII (MPS7; MIM 253220).Identifiants chimiques
| Ac2-223; adipose storage deficiency; AI747421; asd; beta glucuronidase; beta-D-glucuronidase; beta-G1; Beta-glucuronidase; BG; g; glucuronidase beta; glucuronidase, beta; Gur; Gus; GUSB; Gus-r; Gus-s; Gus-t; Gus-u; Gut; MPS7 | |
| 0.18 ng/mL | |
| ELISA | |
| Colorimetric Microplate Reader | |
| GUSB | |
| <10 | |
| HRP | |
| Plasma 50 μL, Serum 50 μL, Supernatant 50 μL | |
| GUSB | |
| 4 hrs 45 mins |
| 0.18 to 40 ng/mL | |
| HRP | |
| Plasma, Serum, Supernatant | |
| 2990 | |
| <12% | |
| Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers | |
| RUO | |
| 2°C to 8°C | |
| 1 hrs 20 mins |
Spécifications
| Ac2-223; adipose storage deficiency; AI747421; asd; beta glucuronidase; beta-D-glucuronidase; beta-G1; Beta-glucuronidase; BG; g; glucuronidase beta; glucuronidase, beta; Gur; Gus; GUSB; Gus-r; Gus-s; Gus-t; Gus-u; Gut; MPS7 | |
| 0.18 ng/mL | |
| ELISA | |
| Colorimetric Microplate Reader | |
| GUSB | |
| <10 | |
| HRP | |
| RUO | |
| 2°C to 8°C | |
| 1 hrs 20 mins |
| 0.18 to 40 ng/mL | |
| HRP | |
| Plasma, Serum, Supernatant | |
| 2990 | |
| <12% | |
| Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers | |
| 96 Tests | |
| Plasma 50 μL, Serum 50 μL, Supernatant 50 μL | |
| GUSB | |
| 4 hrs 45 mins |