missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva HiTrap™ Capto™ PlasmidSelect
For the purification of supercoiled DNA to high quality for gene therapy and DNA vaccine applications
$831.13 - $856.84
Spécifications
| For Use With (Application) | Purification of Supercoiled DNA From Research Scale to cGMP Production |
|---|---|
| Product Type | Chromatography Resin |
| Max. Pressure | 5 bar (0.5 MPa) |
| Matrix | Capto™ PlasmidSelect |
| Length (Metric) | 2.5 cm |
| Numéro de catalogue | Numéro du manufacturier. | Bed Volume (Metric) | Prix | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Numéro de catalogue | Numéro du manufacturier. | Bed Volume (Metric) | Prix | Quantity & Availability | |||||
|
09928117
|
Cytiva
17549911 |
1 mL |
chaque for $831.13
|
|
Connectez-vous ou enregistrez-vous pour vérifier votre prix et la disponibilité.
|
||||
|
09928118
|
Cytiva
17549912 |
5 mL |
chaque for $856.84
|
|
Connectez-vous ou enregistrez-vous pour vérifier votre prix et la disponibilité.
|
||||
Description
- High-flow purification of supercoiled DNA from research scale to cGMP production
- Flexible process design due to a large operational window of flow velocities and bed heights
- Higher throughput for improved productivity and process economy
- Hydrophilic properties of base matrix prevent non-specific binding
- BioProcess resin supported for industrial applications
- Convenient small-scale purification, process development, and scale-up using prepacked HiTrap™ columns
- Capto™ PlasmidSelect is based on a rigid base matrix, delivering excellent pressure-flow properties to plasmid production
Spécifications
| Purification of Supercoiled DNA From Research Scale to cGMP Production | |
| 5 bar (0.5 MPa) | |
| 2.5 cm |
| Chromatography Resin | |
| Capto™ PlasmidSelect |