missing translation for 'onlineSavingsMsg'
Learn More

Cytiva HiTrap™ Capto™ PlasmidSelect

Catalog No. 09928118
Change view
Click to view available options
Bed Volume (Metric):
1 mL
5 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Bed Volume (Metric)
09928118 5 mL
09928117 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 09928118 Supplier Cytiva Supplier No. 17549912
NLSU: Get 1 Consumable Product When You Buy 1 Obtenez 1 produit consommable à l’achat d’un produit
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

For the purification of supercoiled DNA to high quality for gene therapy and DNA vaccine applications

  • High-flow purification of supercoiled DNA from research scale to cGMP production
  • Flexible process design due to a large operational window of flow velocities and bed heights
  • Higher throughput for improved productivity and process economy
  • Hydrophilic properties of base matrix prevent non-specific binding
  • BioProcess resin supported for industrial applications
  • Convenient small-scale purification, process development, and scale-up using prepacked HiTrap™ columns
  • Capto™ PlasmidSelect is based on a rigid base matrix, delivering excellent pressure-flow properties to plasmid production

Specifications

For Use With (Application) Purification of Supercoiled DNA From Research Scale to cGMP Production
Product Type Chromatography Resin
Flow Rate <20 mL/min (Max)
Max. Pressure 5 bar (0.5 MPa)
Matrix Capto™ PlasmidSelect
Bed Volume (Metric) 5 mL
Packaging Quantity 1/Pk.
Diameter (Metric) 1.6 cm
Length (Metric) 2.5 cm