Learn More
Epredia™ HistoStar™ Embedding Workstation Accessories
Numéro de catalogue. p-7220818
Change view
Click to view available options
Product Type:
Attachable Magnifier Assembly
Cold Plate
Embedding Module
Foot Switch
Spatula Kit
Tamping Tool Kit
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Product Type
A81000005 Cold Plate
A81000007 Embedding Module
A81010031 Spatula Kit
A81010030 Tamping Tool Kit
AP16629 Foot Switch
A81010032 Attachable Magnifier Assembly
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Numéro de catalogue. A81000005 Fournisseur Epredia™ Code fournisseur A81000005
Pour de laide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.
Enhance the performance of the Epredia™ HistoStar™ Embedding Workstation with these accessories.
Today's embedding centers can be uncomfortable, hot, or poorly illuminated, leading to fatigue and potential errors. The new Epredia HistoStar Embedding Workstation allows users to embed comfortably and efficiently. With brightly illuminated work surfaces, cool and contoured arm rests and high throughput capacity, this workstation delivers significant improvements in ergonomics and efficiency.
Brightly illuminated and clutter free
  • LED lighting uniformly illuminates the workspace without the clutter of awkward remote lamps
  • A properly illuminated workspace reduces fatigue and minimizes errors
  • Lighting intensity is easily user adjustable
Cool and contoured
  • All user contact points are smooth and insulated, providing a cool and comfortable workspace
  • Pressure points and uncomfortable heat are eliminated
High capacity for high efficiency
  • The capacity of the HistoStar can keep up with even the busiest laboratories
  • Capacity will never limit your workflow with 5 liter paraffin capacity, coldplate area for 72 base molds, large heated workspace and specimen holding area
Any sample size handled with ease
  • The HistoStar offers the first adjustable paraffin dispense paddle
  • A simple adjustment moves the paddle to the most ergonomic position for the user
  • Even Super Mega Cassettes can be embedded with ease
Integrated ParaTrimmer
  • The HistoStar is the only workstation with a heated wax trimmer built directly into the workspace
  • Excess paraffin can be removed at the embedding station, eliminating the need for additional equipment or wax scrapping at the microtome
Intuitive User Interface
  • Large, easy-to-read touchscreen display allows quick access to the temperature controls and other parameters
  • Programmable asleep mode saves energy around your workflow
Dependable Performance
  • The robust HistoStar design minimizes moving parts, reducing wear and service requirements
  • Ensures dependable performance for years to come
Spécifications
For Use With (Equipment) HistoStar Embedding Center
Product Type Cold Plate