missing translation for 'onlineSavingsMsg'
Learn More

Cytiva HiScreen™ Capto™ PlasmidSelect

Catalog No. 09928119
Change view
Click to view available options
Bed Volume (Metric):
4.7 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Bed Volume (Metric)
09928119 4.7 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 09928119 Supplier Cytiva Supplier No. 29201790
NLSU: Get 1 Consumable Product When You Buy 1 Obtenez 1 produit consommable à l’achat d’un produit
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

For purification of supercoiled DNA to high quality, for gene therapy and DNA vaccine applications

  • Allows supercoiled covalently closed circular forms of plasmid DNA to be separated from open circular forms
  • High-flow purification of supercoiled DNA from research scale to cGMP production
  • Flexible process design due to a large operational window of flow velocities and bed heights
  • Convenient small-scale purification, process development, and scale-up using prepacked HiScreen columns
  • Capto™ PlasmidSelect is based on a rigid base matrix, delivering excellent pressure-flow properties to plasmid production
  • Higher throughput for improved productivity and process economy
  • Hydrophilic properties of base matrix prevent non-specific binding
  • BioProcess resin supported for industrial applications

Specifications

For Use With (Application) Gene Therapy and DNA Vaccine Applications
Product Line HiScreen
Product Type Prepacked Column
Diameter (Metric) Inner 0.77 mm
Flow Rate <2.3 mL/min (Max)
Height (Metric) Bed 10 cm
Max. Pressure 8 bar (0.8 MPa)
Matrix Capto™ PlasmidSelect
Bed Volume (Metric) 4.7 mL