Learn More
Cytiva HiScreen™ Capto™ Core 400
Designed for intermediate purification and polishing of viruses and other large biomolecules
Supplier:  Cytiva 17372410
Catalog No. 09928116
Only null left
Explore available promotions
Description
Description
  • For intermediate purification and polishing of viruses and other large biomolecules (Mr > 350,000) in flowthrough mode
  • Offers dual functionality, size exclusion, and binding chromatography
  • An excellent alternative to size‐exclusion chromatography (SEC) resins that are typically employed in the final stages of virus purification in vaccine manufacture
  • Novel core bead technology allows efficient capture of contaminants, while target molecule is collected in the flowthrough
  • Convenient small-scale purification, process development, and scale-up
  • Straightforward optimization due to flowthrough chromatography and robust performance
  • Significantly improved productivity and higher flow rates compared with gel filtration methods
Specifications
Specifications
7.7 mm
Intermediate Purification and Polishing of Viruses, Large Biomolecules
8 bar (0.8 Mpa)
4.7 mL
Capto™ Core 400
0.8 to 3.9 mL/min
100 mm
1/Pk.
7.7 x 100 mm
HiScreen™
Product Suggestions
Product Suggestions