missing translation for 'onlineSavingsMsg'
Learn More

Cytiva HiPrep™ Sephacryl™ S-200 HR Columns

Maximum recovery and minimum nonspecific adsorption

$1392.68 - $1833.15

Specifications

Quantity Each
Particle Size 25 to 75 μm
For Use With (Equipment) Low to medium pressure AKTA™ systems
Product Type Sephacryl HR Column
For Use With (Application) Separation of biomolecules with maximum recovery and minimum nonspecific adsorption with long term chemical and physical stability resulting from the hydrophilic, rigid Allyl Dextran/Bisacrylamide matrix
View More Specs

Products 2
Catalog Number Mfr. No. Flow Rate Bed Volume Diameter (Metric) Inner Price Quantity  
Catalog Number Mfr. No. Flow Rate Bed Volume Diameter (Metric) Inner Price Quantity  
45-000-196
View Documents
Cytiva
17116601
<0.5 mL/min. 120 mL 1.6cm
Each for $1,392.68
Only null left
 
45-000-207
View Documents
Cytiva
17119501
<1.3 mL/min. 320 mL 2.6cm
Each for $1,833.15
Only null left
 
Description

Description

  • Long-term chemical and physical stability from hydrophilic, rigid allyl dextran/bisacrylamide matrix
  • Recommended linear flowrate 5 to 15cm/hr.

Designed for purifying antibodies, serum proteins and midsize proteins

Specifications

Specifications

Each
Low to medium pressure AKTA™ systems
Separation of biomolecules with maximum recovery and minimum nonspecific adsorption with long term chemical and physical stability resulting from the hydrophilic, rigid Allyl Dextran/Bisacrylamide matrix
60cm
1.5 bar [0.15 MPa] (22 psi)
Allyl Dextran and N, N'-methylenebisacrylamide
4°C to 30°C, 20% Ethanol
25 to 75 μm
Sephacryl HR Column
23.62 in.
Polypropylene
3 to 11
Antibodies, serum proteins and mid-size proteins
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'