Learn More
National Safety Apparel™ High Energy Arc Goggles
Catalog No. 17001703
Encompass
Change view
Click to view available options
Color: Black
Catalog No.
17001703 Color: Black
1 options
Catalog No. 17001703 Supplier National Safety Apparel™ Supplier No. H10GGLNN
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Protective goggles with non-vented eyewear venting
  • Anti-fog lens coating
  • Not rated for dust or splash
  • Silicone strap material
  • Polycarbonate lens
Specifications
Size Medium
Material Plastic
Color Black
Seam Type Stitched
Arc Flash Rating 53 cal/cm²
Certifications/Compliance CAT 4, NFPA 70 E, ASTM F2178, and Z87+