missing translation for 'onlineSavingsMsg'
Learn More

Heptafluorobutyric Acid 99.0+%, TCI America™

Catalog No. Q005425G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
Q005425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. Q005425G Supplier TCI America Supplier No. Q005425G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 375-22-4
Molecular Formula C4HF7O2
Molecular Weight (g/mol) 214.039
MDL Number MFCD00004171
InChI Key YPJUNDFVDDCYIH-UHFFFAOYSA-N
Synonym heptafluorobutyric acid, perfluorobutyric acid, heptafluorobutanoic acid, perfluorobutanoic acid, butanoic acid, heptafluoro, heptafluoro-1-butanoic acid, perfluoropropanecarboxylic acid, butyric acid, heptafluoro, kyselina heptafluormaselna, kyselina heptafluormaselna czech
PubChem CID 9777
ChEBI CHEBI:39426
IUPAC Name 2,2,3,3,4,4,4-heptafluorobutanoic acid
SMILES C(=O)(C(C(C(F)(F)F)(F)F)(F)F)O

Specifications

Melting Point -20°C
Color Colorless
Boiling Point 120°C
Quantity 25 g
UN Number 3265
Formula Weight 214.04
Percent Purity ≥99.0% (T)
Physical Form Liquid
Chemical Name or Material Heptafluorobutyric Acid [Sequenation Reagent]
EINECSNumber (2)-1182
RTECSNumber ET4025000
TSCA Yes