missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HEPES Free Acid, >99%, Molecular Biology Reagent, MP Biomedicals™
Supplier: MP Biomedicals Inc 0219482725
Description
Buffering agentSpecifications
| HEPES, free acid | |
| 7365-45-9 | |
| Powder | |
| >99% | |
| MFCD00006158 | |
| JKMHFZQWWAIEOD-UHFFFAOYSA-N | |
| 2-[4-(2-hydroxyethyl)piperazin-1-yl]ethane-1-sulfonic acid | |
| 23831 | |
| 238.3 |
| Free Acid | |
| White | |
| 25 g | |
| C8H18N2O4S | |
| N-2-Hydroxyethylpiperazine-N'-2-ethanesulfonic acid, N-(2-Hydroxyethyl)piperazine-N'-(2-ethanesulfonic acid) | |
| OCCN1CCN(CCS(O)(=O)=O)CC1 | |
| 238.30 | |
| CHEBI:42334 | |
| Molecular Biology Reagent |
Chemical Identifiers
| 7365-45-9 | |
| 238.30 | |
| JKMHFZQWWAIEOD-UHFFFAOYSA-N | |
| 23831 | |
| OCCN1CCN(CCS(O)(=O)=O)CC1 |
| C8H18N2O4S | |
| MFCD00006158 | |
| N-2-Hydroxyethylpiperazine-N'-2-ethanesulfonic acid, N-(2-Hydroxyethyl)piperazine-N'-(2-ethanesulfonic acid) | |
| CHEBI:42334 |
Safety and Handling
EINECSNumber : 230-907-9
storageNote1 : Store desiccated.
Recommended Storage : Store at Room Temperature (15° to 30°C).