missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HEPES Free Acid, >99%, Molecular Biology Reagent, MP Biomedicals™
$100.88 - $812.60
Chemical Identifiers
| CAS | 7365-45-9 |
|---|---|
| Molecular Formula | C8H18N2O4S |
| Molecular Weight (g/mol) | 238.30 |
| MDL Number | MFCD00006158 |
| InChI Key | JKMHFZQWWAIEOD-UHFFFAOYSA-N |
| Synonym | N-2-Hydroxyethylpiperazine-N'-2-ethanesulfonic acid, N-(2-Hydroxyethyl)piperazine-N'-(2-ethanesulfonic acid) |
| PubChem CID | 23831 |
| ChEBI | CHEBI:42334 |
| SMILES | OCCN1CCN(CCS(O)(=O)=O)CC1 |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
ICN19482725
|
MP Biomedicals Inc
0219482725 |
25 g |
Each for $100.88
|
|
|||||
|
ICN19482780
|
MP Biomedicals Inc
0219482780 |
100 g |
Each for $197.04
|
|
|||||
|
ICN19482790
|
MP Biomedicals Inc
0219482790 |
500 g |
Each for $812.60
|
|
|||||
Description
Buffering agentChemical Identifiers
| 7365-45-9 | |
| 238.30 | |
| JKMHFZQWWAIEOD-UHFFFAOYSA-N | |
| 23831 | |
| OCCN1CCN(CCS(O)(=O)=O)CC1 |
| C8H18N2O4S | |
| MFCD00006158 | |
| N-2-Hydroxyethylpiperazine-N'-2-ethanesulfonic acid, N-(2-Hydroxyethyl)piperazine-N'-(2-ethanesulfonic acid) | |
| CHEBI:42334 |
Specifications
| 7365-45-9 | |
| Powder | |
| >99% | |
| MFCD00006158 | |
| JKMHFZQWWAIEOD-UHFFFAOYSA-N | |
| 2-[4-(2-hydroxyethyl)piperazin-1-yl]ethane-1-sulfonic acid | |
| 23831 | |
| 238.3 | |
| HEPES, free acid, Free Acid |
| White | |
| 25 g | |
| C8H18N2O4S | |
| N-2-Hydroxyethylpiperazine-N'-2-ethanesulfonic acid, N-(2-Hydroxyethyl)piperazine-N'-(2-ethanesulfonic acid) | |
| OCCN1CCN(CCS(O)(=O)=O)CC1 | |
| 238.30 | |
| CHEBI:42334 | |
| Molecular Biology Reagent |
Safety and Handling
EINECSNumber : 230-907-9
storageNote1 : Store desiccated.
Recommended Storage : Store at Room Temperature (15° to 30°C).