missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HEPES, Biological Buffer, 99%, Spectrum™ Chemical
$210.77 - $24474.23
Chemical Identifiers
| CAS | 7365-45-9 |
|---|---|
| Molecular Formula | C8H18N2O4S |
| Molecular Weight (g/mol) | 238.30 |
| MDL Number | MFCD00006158 |
| InChI Key | JKMHFZQWWAIEOD-UHFFFAOYSA-N |
| IUPAC Name | 2-[4-(2-hydroxyethyl)piperazin-1-yl]ethane-1-sulfonic acid |
| SMILES | OCCN1CCN(CCS(O)(=O)=O)CC1 |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
18610778
|
Spectrum Chemical Mfg Cor
HE15525GM |
25 g | Poly Bottle |
Each for $210.77
|
|
||||
|
18610776
|
Spectrum Chemical Mfg Cor
HE155100GM |
100 g | Poly Bottle |
Each for $579.17
|
|
||||
|
18610779
|
Spectrum Chemical Mfg Cor
HE155250GM |
250 g | Poly Bottle |
Each for $890.53
|
|
||||
|
18610781
|
Spectrum Chemical Mfg Cor
HE155500GM |
500 g | Poly Bottle |
Each for $1,372.71
|
|
||||
|
18610775
|
Spectrum Chemical Mfg Cor
HE1551KG |
1 kg | Poly Bottle |
Each for $2,660.93
|
|
||||
|
18610780
|
Spectrum Chemical Mfg Cor
HE1555KGBL |
5 kg | Poly Bottle |
Each for $9,815.29
|
|
||||
|
18610777
|
Spectrum Chemical Mfg Cor
HE15512KGBL |
12 kg | Poly Bottle |
N/A
|
N/A | ||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Chemical Identifiers
| 7365-45-9 | |
| 238.30 | |
| JKMHFZQWWAIEOD-UHFFFAOYSA-N | |
| OCCN1CCN(CCS(O)(=O)=O)CC1 |
| C8H18N2O4S | |
| MFCD00006158 | |
| 2-[4-(2-hydroxyethyl)piperazin-1-yl]ethane-1-sulfonic acid |
Specifications
| 7365-45-9 | |
| Pharma and Biopharma | |
| 25 g | |
| C8H18N2O4S | |
| Poly Bottle | |
| OCCN1CCN(CCS(O)(=O)=O)CC1 | |
| 238.30 | |
| Reagent |
| 1 | |
| Crystalline | |
| 0.005 | |
| MFCD00006158 | |
| JKMHFZQWWAIEOD-UHFFFAOYSA-N | |
| 2-[4-(2-hydroxyethyl)piperazin-1-yl]ethane-1-sulfonic acid | |
| 99% | |
| 4-(2-Hydroxyethyl)-1-Piperazineethanesulfonic Acid |
CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.