missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HEPES, Biological Buffer, 99%, Spectrum™ Chemical
Supplier: Spectrum Chemical Mfg Cor HE15512KGBL
Specifications
| 4-(2-Hydroxyethyl)-1-Piperazineethanesulfonic Acid | |
| 1 | |
| Crystalline | |
| 0.005 | |
| MFCD00006158 | |
| JKMHFZQWWAIEOD-UHFFFAOYSA-N | |
| 2-[4-(2-hydroxyethyl)piperazin-1-yl]ethane-1-sulfonic acid | |
| 99% |
| 7365-45-9 | |
| Pharma and Biopharma | |
| 12 kg | |
| C8H18N2O4S | |
| Poly Bottle | |
| OCCN1CCN(CCS(O)(=O)=O)CC1 | |
| 238.30 | |
| Reagent |
Chemical Identifiers
| 7365-45-9 | |
| 238.30 | |
| JKMHFZQWWAIEOD-UHFFFAOYSA-N | |
| OCCN1CCN(CCS(O)(=O)=O)CC1 |
| C8H18N2O4S | |
| MFCD00006158 | |
| 2-[4-(2-hydroxyethyl)piperazin-1-yl]ethane-1-sulfonic acid |
CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.