missing translation for 'onlineSavingsMsg'
Learn More

HEPES, >98%, MP Biomedicals™

$63.12 - $1113.49

Chemical Identifiers

CAS 7365-45-9
Molecular Formula C8H18N2O4S
Molecular Weight (g/mol) 238.30
MDL Number MFCD00006158
InChI Key JKMHFZQWWAIEOD-UHFFFAOYSA-N
Synonym 4-(2-Hydroxyethyl)piperazine-1-ethanesulfonic acid sodium salt, N-(2-Hydroxyethyl)piperazine-N?-(2-ethanesulfonic acid) sodium salt
PubChem CID 23831
ChEBI CHEBI:42334
SMILES OCCN1CCN(CCS(O)(=O)=O)CC1
View More Specs

Products 7
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
ICN19454910
View Documents
MP Biomedicals Inc
0219454910
10 g
Each for $63.12
Only null left
 
ICN19454925
View Documents
MP Biomedicals Inc
0219454925
25 g
Each for $80.69
Only null left
 
ICN19454950
View Documents
MP Biomedicals Inc
0219454950
50 g
Each for $125.73
Only null left
 
ICN19454980
View Documents
MP Biomedicals Inc
0219454980
100 g
Each for $158.53
Only null left
 
ICN19454983
View Documents
MP Biomedicals Inc
0219454983
250 g
Each for $359.53
Only null left
 
ICN19454990
View Documents
MP Biomedicals Inc
0219454990
500 g
Each for $580.82
Only null left
 
ICN19454991
View Documents
MP Biomedicals Inc
0219454991
1 kg
Each for $1,113.49
Only null left
 
Description

Description

Specifications

Chemical Identifiers

7365-45-9
238.30
JKMHFZQWWAIEOD-UHFFFAOYSA-N
23831
OCCN1CCN(CCS(O)(=O)=O)CC1
C8H18N2O4S
MFCD00006158
4-(2-Hydroxyethyl)piperazine-1-ethanesulfonic acid sodium salt, N-(2-Hydroxyethyl)piperazine-N?-(2-ethanesulfonic acid) sodium salt
CHEBI:42334

Specifications

7365-45-9
Powder
C8H18N2O4S
4-(2-Hydroxyethyl)piperazine-1-ethanesulfonic acid sodium salt, N-(2-Hydroxyethyl)piperazine-N?-(2-ethanesulfonic acid) sodium salt
OCCN1CCN(CCS(O)(=O)=O)CC1
238.30
CHEBI:42334
>98%
White
10 g
MFCD00006158
JKMHFZQWWAIEOD-UHFFFAOYSA-N
2-[4-(2-hydroxyethyl)piperazin-1-yl]ethanesulfonic acid
23831
238.3
HEPES
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 230-907-9

Recommended Storage : Store at Room Temperature (15 to 30°C). Store Desiccated.

SDS
missing translation for 'documents'

missing translation for 'documents'