missing translation for 'onlineSavingsMsg'
Learn More

HEPES, >98%, MP Biomedicals™

Supplier:  MP Biomedicals Inc 0219454950

 View more versions of this product

Catalog No. ICN19454950


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

HEPES
White
50 g
MFCD00006158
JKMHFZQWWAIEOD-UHFFFAOYSA-N
2-[4-(2-hydroxyethyl)piperazin-1-yl]ethane-1-sulfonic acid
23831
238.3
7365-45-9
Powder
C8H18N2O4S
4-(2-Hydroxyethyl)piperazine-1-ethanesulfonic acid sodium salt, N-(2-Hydroxyethyl)piperazine-N?-(2-ethanesulfonic acid) sodium salt
OCCN1CCN(CCS(O)(=O)=O)CC1
238.30
CHEBI:42334
>98%

Chemical Identifiers

7365-45-9
238.30
JKMHFZQWWAIEOD-UHFFFAOYSA-N
23831
OCCN1CCN(CCS(O)(=O)=O)CC1
C8H18N2O4S
MFCD00006158
4-(2-Hydroxyethyl)piperazine-1-ethanesulfonic acid sodium salt, N-(2-Hydroxyethyl)piperazine-N?-(2-ethanesulfonic acid) sodium salt
CHEBI:42334
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : 230-907-9

Recommended Storage : Store at Room Temperature (15 to 30°C). Store Desiccated.

SDS
Documents

Documents

Promotions

Promotions