Learn More
Thermo Scientific Chemicals HEPES, 0.5M buffer soln., pH 7.6
CAS: 7365-45-9 | C8H18N2O4S | 238.30 g/mol
Supplier: Thermo Scientific Chemicals J61047AE
Description
HEPES is typically used in cell culture at concentration between 5mM to 30 mM. It is used in cell culture media, in biopharma buffer formulations (both upstream and downstream), and in diagnostic reagents. HEPES based buffers are used in purification bioprocesses of antibodies, bioactives, proteins and growth factors.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsHEPES is typically used in cell culture at concentration between 5mM to 30 mM. It is used in cell culture media, in biopharma buffer formulations (both upstream and downstream), and in diagnostic reagents. HEPES based buffers are used in purification bioprocesses of antibodies, bioactives, proteins and growth factors.
Solubility
Fully miscible in water.
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
Spécifications
| HEPES buffer soln. | |
| 0.5 M | |
| Liquid | |
| C8H18N2O4S | |
| Fully miscible in water. | |
| OCCN1CCN(CCS(O)(=O)=O)CC1 | |
| 238.30 | |
| CHEBI:42334 |
| 7365-45-9 | |
| 7.6 | |
| 100 mL | |
| MFCD00006158 | |
| JKMHFZQWWAIEOD-UHFFFAOYSA-N | |
| 2-[4-(2-hydroxyethyl)piperazin-1-yl]ethanesulfonic acid | |
| 23831 |
Identifiants chimiques
| 7365-45-9 | |
| 238.30 | |
| JKMHFZQWWAIEOD-UHFFFAOYSA-N | |
| CHEBI:42334 |
| C8H18N2O4S | |
| MFCD00006158 | |
| 23831 | |
| OCCN1CCN(CCS(O)(=O)=O)CC1 |
Sécurité et manipulation
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only