missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals HEPES, 0.5M buffer soln., pH 7.6

Catalog No. p-7042012
Change view
Click to view available options
Quantity:
100 mL
250 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ61047AE 100 mL
AAJ61047AK 250 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ61047AE Supplier Thermo Scientific Chemicals Supplier No. J61047AE
Only null left

CAS: 7365-45-9 | C8H18N2O4S | 238.30 g/mol

HEPES is typically used in cell culture at concentration between 5mM to 30 mM. It is used in cell culture media, in biopharma buffer formulations (both upstream and downstream), and in diagnostic reagents. HEPES based buffers are used in purification bioprocesses of antibodies, bioactives, proteins and growth factors.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
HEPES is typically used in cell culture at concentration between 5mM to 30 mM. It is used in cell culture media, in biopharma buffer formulations (both upstream and downstream), and in diagnostic reagents. HEPES based buffers are used in purification bioprocesses of antibodies, bioactives, proteins and growth factors.

Solubility
Fully miscible in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 7365-45-9
Molecular Formula C8H18N2O4S
Molecular Weight (g/mol) 238.30
MDL Number MFCD00006158
InChI Key JKMHFZQWWAIEOD-UHFFFAOYSA-N
PubChem CID 23831
ChEBI CHEBI:42334
SMILES OCCN1CCN(CCS(O)(=O)=O)CC1

Specifications

Concentration 0.5 M
pH 7.6
Physical Form Liquid
Quantity 100 mL
Solubility Information Fully miscible in water.
IUPAC Name 2-[4-(2-hydroxyethyl)piperazin-1-yl]ethanesulfonic acid
Chemical Name or Material HEPES buffer soln.
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only