Learn More
Heidolph™ Heat-On™ Multi-Well Holder Insert
GREENER_CHOICE
Insert for Heidolph Heat-On Multi-Well Holders when used with Hei-PLATE magnetic stirring hotplates.
Supplier:  Heidolph™ 015882280
 Afficher plus de versions de ce produit
Numéro de catalogue. 05404184
Il en reste null
Description
Description
  • Inserts are available for 10 mL, 25 mL, 50 mL, 100 mL and 150 mL flasks
  • Flask inserts also feature cut-away sides for use with two or three neck flasks
  • Multi-tube inserts are available for 16 mm, 20 mm, and 24 mm tubes
  • Available with fluoropolymer coating or anodized finish
Spécifications
Identifiants chimiques
Heat-On Insert
Spécifications
Multi-Well Holder, Hei Series Hotplate
Heat-On Insert
Aluminum
4 x 20 mm Tubes
Anodized
missing translation for 'recommendations'
missing translation for 'recommendations'
Videos
FDS