Learn More
Heidolph™ Heat-On™ Multi-Well Holder Insert
GREENER_CHOICE
Insert for Heidolph Heat-On Multi-Well Holders when used with Hei-PLATE magnetic stirring hotplates.
$290.43 - $602.28
Specifications
Type Heat-On Insert
View More Specs
Products 15
Catalog Number Mfr. No. Coating Holds Price Quantity & Availability  
Catalog Number Mfr. No. Coating Holds Price Quantity & Availability  
05404173
GREENER_CHOICE
View Documents Promotion Details
Heidolph™
015882170
Anodized 10 mL Flasks
Each for $303.87
Only null left
05404157
GREENER_CHOICE
View Documents Promotion Details
Heidolph™
015882010
Fluoropolymer 10 mL Flasks
Each for $305.69
Only null left
05404174
GREENER_CHOICE
View Documents Promotion Details
Heidolph™
015882180
25 mL
Each for $303.87
Only null left
05404158
GREENER_CHOICE
View Documents Promotion Details
Heidolph™
015882020
Fluoropolymer 25 mL Flasks
Each for $295.52
Only null left
05404175
GREENER_CHOICE
View Documents Promotion Details
Heidolph™
015882190
Anodized 50 mL Flasks
Each for $290.43
Only null left
05404159
GREENER_CHOICE
View Documents Promotion Details
Heidolph™
015882030
Fluoropolymer 50 mL Flasks
Each for $295.52
Only null left
05404160
GREENER_CHOICE
View Documents Promotion Details
Heidolph™
015882040
Fluoropolymer 100 mL Flasks
Each for $351.56
Only null left
05404177
GREENER_CHOICE
View Documents Promotion Details
Heidolph™
015882210
150 mL
Each for $488.33
Only null left
05404161
GREENER_CHOICE
View Documents Promotion Details
Heidolph™
015882050
Fluoropolymer 150 mL Flasks
Each for $504.39
Only null left
05404184
GREENER_CHOICE
View Documents Promotion Details
Heidolph™
015882280
Anodized 4 x 20 mm Tubes
Each for $396.11
Only null left
05404168
GREENER_CHOICE
View Documents Promotion Details
Heidolph™
015882120
Fluoropolymer 4 x 20 mm Tubes
Each for $466.63
Only null left
05404185
GREENER_CHOICE
View Documents Promotion Details
Heidolph™
015882290
Anodized 4 x 24 mm Tubes
Each for $396.11
Only null left
05404169
GREENER_CHOICE
View Documents Promotion Details
Heidolph™
015882130
Fluoropolymer 4 x 24 mm Tubes
Each for $466.63
Only null left
05404183
GREENER_CHOICE
View Documents Promotion Details
Heidolph™
015882270
Anodized 8 x 16 mm Tubes
Each for $553.46
Only null left
05404167
GREENER_CHOICE
View Documents Promotion Details
Heidolph™
015882110
Fluoropolymer 8 x 16 mm Tubes
Each for $602.28
Only null left
Description
Description
  • Inserts are available for 10 mL, 25 mL, 50 mL, 100 mL and 150 mL flasks
  • Flask inserts also feature cut-away sides for use with two or three neck flasks
  • Multi-tube inserts are available for 16 mm, 20 mm, and 24 mm tubes
  • Available with fluoropolymer coating or anodized finish
Specifications
Specifications
Heat-On Insert