Learn More
Keystone Safety Heavy-Duty Polypropylene Coveralls, CVL-NW-HD-E
Heavyweight polypropylene coveralls are made of breathable fabric that provides economical protection against dry particulates, dirt, and grime. Available in blue or white.
Supplier:  Keystone Safety CVLNWHDELRGWHITE
 View more versions of this product
Catalog No. 19-055-279
Only null left
Explore available promotions
Description
Description
  • Heavyweight polypropylene coveralls
  • Breathable fabric provides economical protection
  • Elastic wrists and ankles
  • Low cost solution to protect against dry particulates, dirt, and grime
  • 100% latex free
Specifications
Specifications
General Purpose Coveralls
Unisex
White
Elastic
Collared
Serged
Particle
Yes
None
25/Cs.
Large
Polypropylene (Heavy-duty)
Single-use
Elastic
Front Zipper
Non-sterile
Cleanroom Class 7
CVL-NW-HD-E-LRG
Manufacturing, Food Service, Industrial, Medical, Pharmaceutical
Product Suggestions
Product Suggestions