Learn More
Axygen™ Heating Blocks for Microtube Shaker
These nine interchangeable aluminum blocks accommodate PCR plates and a variety of common life science vessels.
Supplier:  Axygen™ I4010A
 Afficher plus de versions de ce produit
Numéro de catalogue. 14223086
Il en reste null
Description
Description
  • No additional tools required for block exchanges
  • Blocks utilize magnetic adhesion technology and are self-recognized once exchanged
  • Combination of heating, cooling, and mixing with versatile block selection makes the Axygen™ Microtube Shaker ideal for many life science laboratories
  • Applications include but are not limited to gene synthesis, gene purification and denaturation, enzymatic reactions, and bacterial growth assays
Spécifications
Identifiants chimiques
Aluminum
Axygen™ Microtube Shaker
Spécifications
1500 rpm
Aluminum
24-well
0.5 mL
Axygen™ Microtube Shaker
missing translation for 'recommendations'
missing translation for 'recommendations'
Videos
FDS