missing translation for 'onlineSavingsMsg'
Learn More

Axygen™ Heating Blocks for Microtube Shaker

Catalog No. p-7227339
Change view
Click to view available options
Tube Size:
0.2 mL
0.5 mL
1.5 mL
2 mL
5 mL
15 mL
50 mL
Speed Range:
1400 rpm
1500 rpm
600 rpm
800 rpm
Well Count:
24-well
4-well
8-well
96-well
96/384-well
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Tube Size Speed Range Well Count
14223092 50 mL 600 rpm 4-well
14223089 5 mL 1500 rpm 8-well
14223090 5 mL 1400 rpm 8-well
14223091 15 mL 800 rpm 8-well
14223086 0.5 mL 1500 rpm 24-well
14223087 1.5 mL 1500 rpm 24-well
14223088 2 mL 1500 rpm 24-well
14223093 0.2 mL 1500 rpm 96-well
14223094 - 1500 rpm 96/384-well
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
Catalog No. 14223092 Supplier Axygen™ Supplier No. I4010G
Only null left

These nine interchangeable aluminum blocks accommodate PCR plates and a variety of common life science vessels.

  • No additional tools required for block exchanges
  • Blocks utilize magnetic adhesion technology and are self-recognized once exchanged
  • Combination of heating, cooling, and mixing with versatile block selection makes the Axygen™ Microtube Shaker ideal for many life science laboratories
  • Applications include but are not limited to gene synthesis, gene purification and denaturation, enzymatic reactions, and bacterial growth assays

Specifications

Speed Range 600 rpm
Tube Size 50 mL
Material Aluminum
For Use With (Equipment) Axygen™ Microtube Shaker
Well Count 4-well