missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Brady™ Hazardous Waste Labels
Hazardous Waste Labels are durable labels that comply with all current Federal regulations regarding identification of waste materials for transportation off-site.
Supplier: Brady™ 60445
Description
- Includes write-on area for adding custom details
- B-235 Coated Paper with pressure sensitive acrylic adhesive from Brady™ adheres to most surfaces
- Labels remove cleanly, leaving no residue on surface
- Operating temperature of -40°F to 150°F (-40°C to 65.5°C)
Identifiants chimiques
| RoHS | |
| Semi-gloss | |
| Paper | |
| Used for general labeling | |
| -40°C to +65°C | |
| 6 in. |
| Permanent Acrylic | |
| Label | |
| Non-sterile | |
| -40°F to 150°F | |
| 0.0038 in. | |
| 152.4 mm |
Spécifications
| RoHS | |
| Black, Red, White | |
| Label | |
| Single-side Printable | |
| Non-sterile | |
| Used for general labeling | |
| -40°F to 150°F | |
| 0.0038 in. | |
| 6 in. | |
| 4 in. |
| Permanent Acrylic | |
| Semi-gloss | |
| Paper | |
| Hazardous Waste Label | |
| Hazardous Waste | |
| 50/Pk. | |
| -40°C to +65°C | |
| 4 x 6 in. | |
| 152.4 mm | |
| 10.1 cm |