missing translation for 'onlineSavingsMsg'
Learn More

Brady™ Hazardous Waste Labels

Hazardous Waste Labels are durable labels that comply with all current Federal regulations regarding identification of waste materials for transportation off-site.

$61.87 - $71.44

Specifications

Certifications/Compliance RoHS
Adhesive Type Permanent Acrylic
Finish Semi-gloss
Format Label
Material Paper
View More Specs

Products 3
Catalog Number Mfr. No. Color Quantity Display Text Height (English) Price Quantity & Availability  
Catalog Number Mfr. No. Color Quantity Display Text Height (English) Price Quantity & Availability  
19054847
View Documents Promotion Details
Brady™
60447
White, Green 100/Pk. Non-Hazardous Waste 6 in.
Pack of 100 Each for $62.51
Only null left
 
19054846
View Documents Promotion Details
Brady™
60446
White, Blue 100/Pk. Non-regulated Waste 6 in.
Pack of 100 Each for $71.44
Only null left
 
19054845
View Documents Promotion Details
Brady™
60445
Black, Red, White 50/Pk. Hazardous Waste 4 in.
Pack of 100 Each for $61.87
Only null left
 
Description

Description

  • Includes write-on area for adding custom details
  • B-235 Coated Paper with pressure sensitive acrylic adhesive from Brady™ adheres to most surfaces
  • Labels remove cleanly, leaving no residue on surface
  • Operating temperature of -40°F to 150°F (-40°C to 65.5°C)
Specifications

Specifications

RoHS
Semi-gloss
Paper
Used for general labeling
-40°C to +65°C
6 in.
Permanent Acrylic
Label
Non-sterile
-40°F to 150°F
0.0038 in.
152.4 mm
Videos
SDS
Documents

Documents