Learn More
Thermo Scientific™ Harvestainer™ Microcarrier Separation System
GREENER_CHOICE
The Thermo Scientific Harvestainer Microcarrier Separation System is the ideal solution for the separation of microcarrier beads during adherent cell culture production.
Supplier:  Thermo Scientific™ SH31071.02
 Afficher plus de versions de ce produit
Numéro de catalogue. SH3107102
Il en reste null
Description
Description
The Thermo Scientific Harvestainer Microcarrier Separation System is the ideal solution for the separation of microcarrier beads during adherent cell culture production. The Harvestainer system is a closed, single-use system that helps to increase product yields versus traditional methods, while reducing clean-in-place and steam-in-place requirements and water for injection usage. It eliminates equipment maintenance and reduces process validations, cycle times, and manual final decanting processes.
For smaller-scale or larger-scale applications, the Harvestainer system is ideal for in-line microcarrier separation. The 3 L and 12 L Harvestainer systems consist of a pre-assembled bioprocess container (BPC) and tray in a single-use unit, where the tray acts as the secondary containment device. The 25 L and 50 L Harvestainer systems consist of a 3D BPC with a drum as the secondary containment device. The BPC features a dip tube with dipwell design for better drainage and minimal manipulation.
• Closed system
• Scalable for both smaller-scale and larger-scale applications
• Helps deliver high recovery rates
• Inline microcarrier separation
Applications
• Adherent cell culture production
Spécifications
Identifiants chimiques
Sterile
Room Temperature
Room Temperature
Harvestainer
Spécifications
Sterile
C-Flex
50 L
Harvestainer
50 L
Room Temperature
Room Temperature
missing translation for 'recommendations'
missing translation for 'recommendations'
Videos
FDS
Documents
Documents
Promotions
Promotions
For Research Use Only. Not for use in diagnostic procedures.