missing translation for 'onlineSavingsMsg'
Learn More

ThermoHeat™ Hard Hat Liners: Polar-Tec Fleece Long-Neck Length

Catalog No. 296361304
Change view
Click to view available options
Color:
Gray
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
296361304 Gray
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 296361304 Supplier ThermoHeat™ Supplier No. 1070025

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Gray with elastic around face for snug fit and protection

  • Includes:
    • Three suspension tabs
    • Velcro chin closure

Specifications

Product Type Cold-weather Hard Hat Liner
Size Universal
Material Fleece
Color Gray
Disposable Reusable
For Use With (Application) Cold Protection Hood