missing translation for 'onlineSavingsMsg'
Learn More

3M™ Hard Hat Front Visor Seal, GR2B-10

Catalog No. 296366629
Change view
Click to view available options
Orange
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
296366629 Orange
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 296366629 Supplier 3M™ Supplier No. 7000103819

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Pliable, water-resistant PVC coated nylon front visor that helps keep water and debris from falling behind faceshield

  • This pliable, water-resistant PVC coated nylon front visor helps keep water and debris from falling behind faceshield
  • 3M provides reliable head and face protection products that meet regulated performance and safety requirements.

Specifications

Color Orange
For Use With (Equipment) 3M Peltor V Series Visors
Product Type Front Visor Seal
Width (Metric) 1.27 cm
Width (English) 0.5 in.