missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Promega GoTaq™ Flexi DNA Polymerase
Taq DNA Polymerase. Provided with Green and Colorless Reaction Buffers without MgCl2, and 25mM MgCl2. Green Buffer allows direct gel analysis after amplification. Enhanced performance over standard Taq DNA Polymerase.
Supplier: Promega M8297
Comprend: GoTaq Flexi DNA Polymerase, 5X Colorless GoTaq Flexi Buffer (Used when direct fluorescence or absorbance readings are required without prior purification of the DNA from the amplification reaction), 5X Green GoTaq Flexi Buffer (Contains two dyes that separate during electrophoresis to facilitate direct-to-gel analysis of PCR products), 25mM MgCl2
Description
- Optimize Enzyme and Magnesium Concentrations in Your PCR
- Mg-free reaction buffers for ease of optimization
- Direct-to-gel convenience
- Supplied at a concentration of 5u/μL
Identifiants chimiques
| 5 U/μL | |
| DNA Polymerase | |
| 8 | |
| GoTaq Flexi DNA Polymerase, 5X Colorless GoTaq Flexi buffer (Used when direct fluorescence or absorbance readings are required without prior purification of the DNA from the amplification reaction), 5X Green GoTaq Flexi buffer, 25mM MgCl2 |
| −30 to −10°C | |
| Liquid | |
| Standard PCR |
Spécifications
| 5 U/μL | |
| DNA Polymerase | |
| 8 | |
| Standard PCR |
| −30 to −10°C | |
| Liquid | |
| 5000U | |
| GoTaq Flexi DNA Polymerase, 5X Colorless GoTaq Flexi buffer (Used when direct fluorescence or absorbance readings are required without prior purification of the DNA from the amplification reaction), 5X Green GoTaq Flexi buffer, 25mM MgCl2 |