missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gold ICP Standard, 1000 ppm Au in 3% HCl, Ricca Chemical
$710.90 - $1,724.83
Chemical Identifiers
| CAS | 7440-57-5 |
|---|---|
| Molecular Formula | Au |
| Molecular Weight (g/mol) | 196.97 |
| MDL Number | MFCD00003436 |
| InChI Key | PCHJSUWPFVWCPO-UHFFFAOYSA-N |
| PubChem CID | 23985 |
| ChEBI | CHEBI:30050 |
| IUPAC Name | gold |
| SMILES | [Au] |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
PAU1KH-100
|
Ricca Chemical Company
PAU1KH100 |
100 mL | Amber Glass Bottle |
N/A
|
|
||||
|
PAU1KH-500
|
Ricca Chemical Company
PAU1KH500 |
500 mL | Amber Glass Bottle |
N/A
|
|
||||
Chemical Identifiers
| 7440-57-5 | |
| 196.97 | |
| PCHJSUWPFVWCPO-UHFFFAOYSA-N | |
| CHEBI:30050 | |
| [Au] |
| Au | |
| MFCD00003436 | |
| 23985 | |
| gold |
Specifications
| Amber Glass Bottle | |
| 1,000ppm Au | |
| 97.0,4.14,0.1 | |
| Liquid | |
| MFCD00003436 | |
| Standard | |
| [Au] | |
| 196.97 | |
| 23985 | |
| Gold ICP Standard |