CAS RN 7440-57-5
Gold standard solution, for AAS, 1 mg/ml Au in 10-20% HCl
CAS: 7440-57-5 | Au | 196.97 g/mol
Gold nanoparticles, 20nm, carboxy-functionalized, 3kDa PEGylated, OD50, 524nm absorption, Thermo Scientific™
CAS: 7440-57-5 Molecular Formula: Au Molecular Weight (g/mol): 196.967 MDL Number: MFCD00003436 InChI Key: PCHJSUWPFVWCPO-UHFFFAOYSA-N Synonym: colloidal,powder,flake,leaf,gold, colloidal,burnish,shell,ci pigment metal 3,magnesium purple,c.i. pigment metal 3 PubChem CID: 23985 ChEBI: CHEBI:30050 IUPAC Name: gold SMILES: [Au]
Gold nanoparticles, 20nm, amine-functionalized, 3kDa PEGylated, OD50, 524nm absorption, Thermo Scientific™
CAS: 7440-57-5 Molecular Formula: Au Molecular Weight (g/mol): 196.967 MDL Number: MFCD00003436 InChI Key: PCHJSUWPFVWCPO-UHFFFAOYSA-N Synonym: colloidal,powder,flake,leaf,gold, colloidal,burnish,shell,ci pigment metal 3,magnesium purple,c.i. pigment metal 3 PubChem CID: 23985 ChEBI: CHEBI:30050 IUPAC Name: gold SMILES: [Au]
Gold wire, 0.127mm (0.005in) dia, Premion™, 99.99% (metals basis)
CAS: 7440-57-5 Molecular Formula: Au Molecular Weight (g/mol): 196.97 MDL Number: MFCD00003436 InChI Key: PCHJSUWPFVWCPO-UHFFFAOYSA-N Synonym: colloidal,powder,flake,leaf,gold, colloidal,burnish,shell,ci pigment metal 3,magnesium purple,c.i. pigment metal 3 PubChem CID: 23985 ChEBI: CHEBI:30050 IUPAC Name: gold SMILES: [Au]
Gold wire, 0.2mm (0.008in) dia, 99.9% (metals basis)
CAS: 7440-57-5 Molecular Formula: Au Molecular Weight (g/mol): 196.97 MDL Number: MFCD00003436 InChI Key: PCHJSUWPFVWCPO-UHFFFAOYSA-N Synonym: colloidal,powder,flake,leaf,gold, colloidal,burnish,shell,ci pigment metal 3,magnesium purple,c.i. pigment metal 3 PubChem CID: 23985 ChEBI: CHEBI:30050 IUPAC Name: gold SMILES: [Au]
Gold splatter, 99.9% (metals basis)
CAS: 7440-57-5 Molecular Formula: Au Molecular Weight (g/mol): 196.97 MDL Number: MFCD00003436 InChI Key: PCHJSUWPFVWCPO-UHFFFAOYSA-N Synonym: colloidal,powder,flake,leaf,gold, colloidal,burnish,shell,ci pigment metal 3,magnesium purple,c.i. pigment metal 3 PubChem CID: 23985 ChEBI: CHEBI:30050 IUPAC Name: gold SMILES: [Au]
Gold flake/sphere, APS 2.2-3.5 micron, 99.96% (metals basis)
CAS: 7440-57-5 Molecular Formula: Au Molecular Weight (g/mol): 196.97 MDL Number: MFCD00003436 InChI Key: PCHJSUWPFVWCPO-UHFFFAOYSA-N Synonym: colloidal,powder,flake,leaf,gold, colloidal,burnish,shell,ci pigment metal 3,magnesium purple,c.i. pigment metal 3 PubChem CID: 23985 ChEBI: CHEBI:30050 IUPAC Name: gold SMILES: [Au]
Gold flake/sphere, APS 1.5-3.0 micron, 99.96% (metals basis), Thermo Scientific™
CAS: 7440-57-5 Molecular Formula: Au Molecular Weight (g/mol): 196.97 MDL Number: MFCD00003436 InChI Key: PCHJSUWPFVWCPO-UHFFFAOYSA-N Synonym: colloidal,powder,flake,leaf,gold, colloidal,burnish,shell,ci pigment metal 3,magnesium purple,c.i. pigment metal 3 PubChem CID: 23985 ChEBI: CHEBI:30050 IUPAC Name: gold SMILES: [Au]
Gold wire, 2.0mm (0.08in) dia, Premion™, 99.999% (metals basis)
CAS: 7440-57-5 Molecular Formula: Au Molecular Weight (g/mol): 196.97 MDL Number: MFCD00003436 InChI Key: PCHJSUWPFVWCPO-UHFFFAOYSA-N Synonym: colloidal,powder,flake,leaf,gold, colloidal,burnish,shell,ci pigment metal 3,magnesium purple,c.i. pigment metal 3 PubChem CID: 23985 ChEBI: CHEBI:30050 IUPAC Name: gold SMILES: [Au]
Gold wire, 2.0mm (0.08in) dia, 99.95% (metals basis)
CAS: 7440-57-5 Molecular Formula: Au Molecular Weight (g/mol): 196.97 MDL Number: MFCD00003436 InChI Key: PCHJSUWPFVWCPO-UHFFFAOYSA-N Synonym: colloidal,powder,flake,leaf,gold, colloidal,burnish,shell,ci pigment metal 3,magnesium purple,c.i. pigment metal 3 PubChem CID: 23985 ChEBI: CHEBI:30050 IUPAC Name: gold SMILES: [Au]
Gold wire, 0.5mm (0.02in) dia, Premion™, 99.999% (metals basis)
CAS: 7440-57-5 Molecular Formula: Au Molecular Weight (g/mol): 196.97 MDL Number: MFCD00003436 InChI Key: PCHJSUWPFVWCPO-UHFFFAOYSA-N Synonym: colloidal,powder,flake,leaf,gold, colloidal,burnish,shell,ci pigment metal 3,magnesium purple,c.i. pigment metal 3 PubChem CID: 23985 ChEBI: CHEBI:30050 IUPAC Name: gold SMILES: [Au]
Gold powder, spherical, APS 5.5-9.0 micron, 99.96+% (metals basis)
CAS: 7440-57-5 Molecular Formula: Au Molecular Weight (g/mol): 196.97 MDL Number: MFCD00003436 InChI Key: PCHJSUWPFVWCPO-UHFFFAOYSA-N Synonym: colloidal,powder,flake,leaf,gold, colloidal,burnish,shell,ci pigment metal 3,magnesium purple,c.i. pigment metal 3 PubChem CID: 23985 ChEBI: CHEBI:30050 IUPAC Name: gold SMILES: [Au]
Gold wire, 1.0mm (0.04in) dia, 99.95% (metals basis)
CAS: 7440-57-5 Molecular Formula: Au Molecular Weight (g/mol): 196.97 MDL Number: MFCD00003436 InChI Key: PCHJSUWPFVWCPO-UHFFFAOYSA-N Synonym: colloidal,powder,flake,leaf,gold, colloidal,burnish,shell,ci pigment metal 3,magnesium purple,c.i. pigment metal 3 PubChem CID: 23985 ChEBI: CHEBI:30050 IUPAC Name: gold SMILES: [Au]
Gold powder, spherical, <10 micron, 99.95% (metals basis)
CAS: 7440-57-5 Molecular Formula: Au Molecular Weight (g/mol): 196.97 MDL Number: MFCD00003436 InChI Key: PCHJSUWPFVWCPO-UHFFFAOYSA-N Synonym: colloidal,powder,flake,leaf,gold, colloidal,burnish,shell,ci pigment metal 3,magnesium purple,c.i. pigment metal 3 PubChem CID: 23985 ChEBI: CHEBI:30050 IUPAC Name: gold SMILES: [Au]
Gold sponge, -20 mesh, 99.95% (metals basis)
CAS: 7440-57-5 Molecular Formula: Au Molecular Weight (g/mol): 196.97 MDL Number: MFCD00003436 InChI Key: PCHJSUWPFVWCPO-UHFFFAOYSA-N Synonym: colloidal,powder,flake,leaf,gold, colloidal,burnish,shell,ci pigment metal 3,magnesium purple,c.i. pigment metal 3 PubChem CID: 23985 ChEBI: CHEBI:30050 IUPAC Name: gold SMILES: [Au]