missing translation for 'onlineSavingsMsg'
Learn More

Gold ICP Standard, 1000 ppm Au in 3% HCl, Ricca Chemical

Numéro de catalogue. PAU1KH100
Change view
Click to view available options
Quantity:
100 mL
500 mL
Packaging:
Amber Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity Packaging
PAU1KH100 100 mL Amber Glass Bottle
PAU1KH500 500 mL Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Numéro de catalogue. PAU1KH100 Fournisseur Ricca Chemical Company Code fournisseur PAU1KH100
Il en reste null

Identifiants chimiques

CAS 7440-57-5 , 7647-01-0 , 7732-18-5
Molecular Formula Au
Molecular Weight (g/mol) 196.97
MDL Number MFCD00003436
InChI Key PCHJSUWPFVWCPO-UHFFFAOYSA-N
PubChem CID 23985
ChEBI CHEBI:30050
IUPAC Name gold
SMILES [Au]

Spécifications

Packaging Amber Glass Bottle
Concentration or Composition Notes 1,000ppm Au
CAS Min % 94.0, 3.98, 0.09
CAS Max % 97.0, 4.14, 0.1
Equivalence 1 mL = 1 mg Au
Physical Form Liquid
Boiling Point 100°C
Type Standard
Quantity 100 mL
Chemical Name or Material Gold ICP Standard