missing translation for 'onlineSavingsMsg'
Learn More

Gold ICP Standard, 10,000 ppm Au in 5% HCl, Ricca Chemical

$2787.90 - $2787.90

Chemical Identifiers

CAS 7440-57-5
Molecular Formula Au
Molecular Weight (g/mol) 196.97
MDL Number MFCD00003436
InChI Key PCHJSUWPFVWCPO-UHFFFAOYSA-N
PubChem CID 23985
ChEBI CHEBI:30050
IUPAC Name gold
SMILES [Au]
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
PAU10KH-100
View Documents
Ricca Chemical Company
PAU10KH100
100 mL Amber Glass Bottle
Each for $2,787.90
Only null left
 
Description

Description

Specifications

Chemical Identifiers

7440-57-5
196.97
PCHJSUWPFVWCPO-UHFFFAOYSA-N
CHEBI:30050
[Au]
Au
MFCD00003436
23985
gold

Specifications

Amber Glass Bottle
10,000ppm Au
87.0,11.25,0.87
Liquid
MFCD00003436
Standard
[Au]
196.97
23985
Gold ICP Standard
7440-57-5 , 7647-01-0 , 7732-18-5
83.0,10.8,0.83
1 mL = 10 mg Au
Au
Approximately 0°C
PCHJSUWPFVWCPO-UHFFFAOYSA-N
gold
100 mL
CHEBI:30050
Videos
SDS
Documents

Documents