missing translation for 'onlineSavingsMsg'
Learn More

Gold ICP Standard, 10,000 ppm Au in 5% HCl, Ricca Chemical

Catalog No. PAU10KH100
Change view
Click to view available options
Quantity:
100 mL
Packaging:
Amber Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
PAU10KH100 100 mL Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PAU10KH100 Supplier Ricca Chemical Company Supplier No. PAU10KH100
Only null left

Chemical Identifiers

CAS 7440-57-5 , 7647-01-0 , 7732-18-5
Molecular Formula Au
Molecular Weight (g/mol) 196.97
MDL Number MFCD00003436
InChI Key PCHJSUWPFVWCPO-UHFFFAOYSA-N
PubChem CID 23985
ChEBI CHEBI:30050
IUPAC Name gold
SMILES [Au]

Specifications

Packaging Amber Glass Bottle
Concentration or Composition Notes 10,000ppm Au
CAS Min % 83.0, 10.8, 0.83
CAS Max % 87.0, 11.25, 0.87
Equivalence 1 mL = 10 mg Au
Physical Form Liquid
Melting Point Approximately 0°C
Type Standard
Quantity 100 mL
Chemical Name or Material Gold ICP Standard