missing translation for 'onlineSavingsMsg'
Learn More

Ergodyne™ GloWear™ 8918BK Class E Black Bottom Rain Bibs

Catalog No. 19820917
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
5X-Large
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19820917 5X-Large
19820901 Small
19820902 Medium
19820903 Large
19820904 X-Large
19820908 2X-Large
19820911 3X-Large
19820916 4X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Catalog No. 19820917 Supplier Ergodyne™ Supplier No. 25019

Class E Black Bottom Rain Bibs provide breathable, waterproof, comfort in a bib style with black accents to hide dirt.

  • Weatherproof 300D oxford outer shell
  • Breathable PU coating
  • 2 in. ANSI-compliant 3M™ Scotchlite™ Reflective
  • Adjustable elastic suspenders
  • Zippered leg openings for easy on/off
  • One right leg cargo pocket
  • Black accents on bottom of leg

Specifications

Size 5X-Large
Material Polyurethane
Color Yellow
Waist Style Adjustable elastic suspenders
For Use With (Application) Flagging Crew, Road Construction, Utility Crew, Survey Crew, Construction