missing translation for 'onlineSavingsMsg'
Learn More

Ergodyne™ GloWear™ 8004 Hi-Vis Seat Belt Cover

Catalog No. 19910169
Change view
Click to view available options
Color:
Lime
Orange
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
19910169 Lime
19910168 Orange
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 19910169 Supplier Ergodyne™ Supplier No. 29043

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

GloWear™ 8004 Hi-Vis Seat Belt Cover enchances the visibility of the driver.

  • ANSI-compliant polyester solid (3.6 oz.)
  • 2 in. ANSI-compliant reflective material
  • Fits standard width seat belt harness

Specifications

Closure Type Hook and Loop
Color Lime
For Use With (Equipment) Seat Belt
Material Polyester