missing translation for 'onlineSavingsMsg'
Learn More

GENTEC™ Genstar Technologies™ Gentec™ M22 Series Metering Valves

Catalog No. 10575165
Change view
Click to view available options
Port Type:
1/4 in. GENLOK End Connections
1/4 in. Male NPT End Connections
1/8 in. GENLOK End Connections
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Port Type
10575165 1/4 in. Male NPT End Connections
10575163 1/8 in. GENLOK End Connections
10575164 1/4 in. GENLOK End Connections
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 10575165 Supplier GENTEC™ Supplier No. SS MV22 NT4

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Includes

Handle, Panel Nut, Bonnet Sleeve, Valve Stem, Bonnet, Lower Gland, O-ring, Body

With maximum operating pressure of 1000 psi (69 bar)

  •  GENLOK and NPT end connections
  • Panel mounting available
  • Cleaned and degreased for oxygen service according to CGA G-4.1 and ASTM G93 Class C, respectively

Specifications

Type Valve
Temperature (English) Max. -10°F to +400°F
Temperature (Metric) Max -23°C to +204°C
Model SS-MV22-NT4
Includes Handle, Panel Nut, Bonnet Sleeve, Valve Stem, Bonnet, Lower Gland, O-ring Fluorine, Body
Port Type 1/4 in. Male NPT End Connections
Material 316 Stainless Steel (Handle, Panel Nut, Bonnet Sleeve, Valve Stem, Bonnet, Body), PTFE (Lower Gland), Fluorine Rubber (O-Ring)
Pressure 1000 psi