missing translation for 'onlineSavingsMsg'
Learn More

Gallium ICP Standard, 10,000 ppm Ga in 5% HNO3, Ricca Chemical

Supplier:  Ricca Chemical Company PGA10KN100

 View more versions of this product

Catalog No. PGA10KN-100


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Gallium ICP Standard
7440-55-3 , 7697-37-2 , 7732-18-5
78.0,23.39,0.78
Ga
Colorless
Standard
[Ga]
69.72
5360835
1mL = 10mg Ga (10, 000ppm Ga)Ga in 5% HNO3
Natural LDPE Bottle
75.0,22.47,0.75
Liquid
MFCD00134045
< 2
GYHNNYVSQQEPJS-UHFFFAOYSA-N
gallium
100 mL
CHEBI:49631

Chemical Identifiers

7440-55-3
69.72
GYHNNYVSQQEPJS-UHFFFAOYSA-N
CHEBI:49631
[Ga]
Ga
MFCD00134045
5360835
gallium
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

ShelfLife : 18 months

SDS
Documents

Documents

Promotions

Promotions